Merck
Syntaxin 8 impairs trafficking of cystic fibrosis transmembrane conductance regulator (CFTR) and inhibits its channel activity.
Journal of cell science (2004-03-25)
Frédéric Bilan, Vincent Thoreau, Magali Nacfer, Renaud Dérand, Caroline Norez, Anne Cantereau, Martine Garcia, Frédéric Becq, Alain Kitzis
RESUMEN
The cystic fibrosis transmembrane conductance regulator (CFTR) is a cyclic AMP-dependent chloride channel that mediates electrolyte transport across the luminal surface of epithelial cells. In this paper, we describe the CFTR regulation by syntaxin 8, a t-SNARE protein (target soluble N-ethylmaleimide-sensitive factor attachment protein receptor) involved in the SNARE endosomal complex. Syntaxin family members are key molecules implicated in diverse vesicle docking and membrane fusion events. We found that syntaxin 8 physically interacts with CFTR: recombinant syntaxin 8 binds CFTR in vitro and both proteins co-immunoprecipitate in HT29 cells. Syntaxin 8 regulates CFTR-mediated currents in chinese hamster ovary (CHO) cells stably expressing CFTR and syntaxin 8. Iodide efflux and whole-cell patch-clamp experiments on these cells indicate a strong inhibition of CFTR chloride current by syntaxin 8 overexpression. At the cellular level, we observed that syntaxin 8 overexpression disturbs CFTR trafficking. Confocal microscopy shows a dramatic decrease in green fluorescent protein-tagged CFTR plasma membrane staining, when syntaxin 8 is coexpressed in COS-7 cells. Using antibodies against Lamp-1, TfR or Rab11 we determined by immunofluorescence assays that both proteins are mainly accumulated in recycling endosomes. Our results evidence that syntaxin 8 contributes to the regulation of CFTR trafficking and chloride channel activity by the SNARE machinery.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-CFTR Antibody, a.a. 1370-1380, clone M3A7, clone M3A7, Chemicon®, from mouse
En este momento no podemos mostrarle ni los precios ni la disponibilidad