Merck
  • In vivo biodistribution of no-carrier-added 6-18F-fluoro-3,4-dihydroxy-L-phenylalanine (18F-DOPA), produced by a new nucleophilic substitution approach, compared with carrier-added 18F-DOPA, prepared by conventional electrophilic substitution.
In vivo biodistribution of no-carrier-added 6-18F-fluoro-3,4-dihydroxy-L-phenylalanine (18F-DOPA), produced by a new nucleophilic substitution approach, compared with carrier-added 18F-DOPA, prepared by conventional electrophilic substitution.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-12-17)
Willem-Jan Kuik, Ido P Kema, Adrienne H Brouwers, Rolf Zijlma, Kiel D Neumann, Rudi A J O Dierckx, Stephen G DiMagno, Philip H Elsinga
RESUMEN
A novel synthetic approach to 6-(18)F-fluoro-3,4-dihydroxy-L-phenylalanine ((18)F-DOPA), involving the nucleophilic substitution of a diaryliodonium salt precursor with non-carrier-added (18)F-fluoride, yielded a product with a specific activity that was 3 orders of magnitude higher than the product of the conventional synthesis method, involving an electrophilic substitution of a trialkylstannane precursor with (18)F2. We performed a direct comparison of high- and low-specific-activity (18)F-DOPA in a neuroendocrine tumor model to determine whether this difference in specific activity has implications for the biologic behavior and imaging properties of (18)F-DOPA. (18)F-DOPA was produced via the novel synthesis method, yielding (18)F-DOPA-H with a high specific activity (35,050 ± 4,000 GBq/mmol). This product was compared in several experiments with conventional (18)F-DOPA-L with a low specific activity (11 ± 2 GBq/mmol). In vitro accumulation experiments with the human pancreatic neuroendocrine tumor cell line BON-1 were performed at both 0 °C and 37 °C and at 37 °C in the presence of pharmacologic inhibitors of proteins involved in the uptake mechanism of (18)F-DOPA. Small-animal PET experiments were performed in athymic nude mice bearing a BON-1 tumor xenograft. At 37 °C, the uptake of both (18)F-DOPA-H and (18)F-DOPA-L did not differ significantly during a 60-min accumulation experiment in BON-1 cells. At 0 °C, the uptake of (18)F-DOPA-L was significantly decreased, whereas the lower temperature did not alter the uptake of (18)F-DOPA-H. The pharmacologic inhibitors carbidopa and tetrabenazine also revealed differential effects between the 2 types of (18)F-DOPA in the 60-min accumulation experiment. The small-animal PET experiments did not show any significant differences in distribution and metabolism of (18)F-DOPA-H and (18)F-DOPA-L in carbidopa-pretreated mice. The advantages of the novel synthesis of (18)F-DOPA, which relies on nucleophilic fluorination of a diaryliodonium salt precursor, lie in the simplicity of the synthesis method, compared with the conventional, electrophilic approach and in the reduced mass of administered, pharmacologically active (19)F-DOPA. (18)F-DOPA-H demonstrated comparable imaging properties in an in vivo model for neuroendocrine tumors, despite the fact that the injected mass of material was 3 orders of magnitude less than (18)F-DOPA-L.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
3,4-Dihydroxy-DL-phenylalanine
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Tantalum(V) ethoxide, packaged for use in deposition systems
En este momento no podemos mostrarle ni los precios ni la disponibilidad