Merck
  • The elusive structure of Pd2(dba)3. Examination by isotopic labeling, NMR spectroscopy, and X-ray diffraction analysis: synthesis and characterization of Pd2(dba-Z)3 complexes.
The elusive structure of Pd2(dba)3. Examination by isotopic labeling, NMR spectroscopy, and X-ray diffraction analysis: synthesis and characterization of Pd2(dba-Z)3 complexes.
Journal of the American Chemical Society (2013-05-25)
Anant R Kapdi, Adrian C Whitwood, David C Williamson, Jason M Lynam, Michael J Burns, Thomas J Williams, Alan J Reay, Jordan Holmes, Ian J S Fairlamb
RESUMEN
Pd(0)2(dba)3 (dba = E,E-dibenzylidene acetone) is the most widely used Pd(0) source in Pd-mediated transformations. Pd(0)2(dba-Z)3 (Z = dba aryl substituents) complexes exhibit remarkable and differential catalytic performance in an eclectic array of cross-coupling reactions. The precise structure of these types of complexes has been confounding, since early studies in 1970s to the present day. In this study the solution and solid-state structures of Pd(0)2(dba)3 and Pd(0)2(dba-Z)3 have been determined. Isotopic labeling ((2)H and (13)C) has allowed the solution structures of the freely exchanging major and minor isomers of Pd(0)2(dba)3 to be determined at high field (700 MHz). DFT calculations support the experimentally determined major and minor isomeric structures, which show that the major isomer of Pd(0)2(dba)3 possesses bridging dba ligands found exclusively in a s-cis,s-trans conformation. For the minor isomer one of the dba ligands is found exclusively in a s-trans,s-trans conformation. Single crystal X-ray diffraction analysis of Pd(0)2(dba)3·CHCl3 (high-quality data) shows that all three dba ligands are found over two positions. NMR spectroscopic analysis of Pd(0)2(dba-Z)3 reveals that the aryl substituent has a profound effect on the rate of Pd-olefin exchange and the global stability of the complexes in solution. Complexes containing the aryl substituents, 4-CF3, 4-F, 4-t-Bu, 4-hexoxy, 4-OMe, exhibit well-resolved (1)H NMR spectra at 298 K, whereas those containing 3,5-OMe and 3,4,5-OMe exhibit broad spectra. The solid-state structures of three Pd(0)2(dba-Z)3 complexes (4-F, 4-OMe, 3,5-OMe) have been determined by single crystal X-ray diffraction methods, which have been compared with Goodson's X-ray structure of Pd(0)2(dba-4-OH)3.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tris(dibencilideniacetona)dipaladio(0), 97%