Merck
PDGF-C: a new performer in the neurovascular interplay.
Trends in molecular medicine (2013-05-30)
Chunsik Lee, Fan Zhang, Zhongshu Tang, Yizhi Liu, Xuri Li
RESUMEN
The importance of neurovascular crosstalk in development, normal physiology, and pathologies is increasingly being recognized. Although vascular endothelial growth factor (VEGF), a prototypic regulator of neurovascular interaction, has been studied intensively, defining other important regulators in this process is warranted. Recent studies have shown that platelet-derived growth factor C (PDGF-C) is both angiogenic and a neuronal survival factor, and it appears to be an important component of neurovascular crosstalk. Importantly, the expression pattern and functional properties of PDGF-C and its receptors differ from those of VEGF, and thus the PDGF-C-mediated neurovascular interaction may represent a new paradigm of neurovascular crosstalk.