Merck
Guanabenz, which enhances the unfolded protein response, ameliorates mutant SOD1-induced amyotrophic lateral sclerosis.
Neurobiology of disease (2014-08-20)
Lijun Wang, Brian Popko, Emily Tixier, Raymond P Roos
RESUMEN
Approximately 20% of familial amyotrophic lateral sclerosis (FALS) cases are caused by mutant superoxide dismutase type 1 (mtSOD1). Although the mechanisms of mtSOD1-induced toxicity remain poorly understood, evidence suggests that accumulation of misfolded SOD1 is fundamental to its toxicity and the death of motor neurons. Misfolded mtSOD1 can accumulate inside the endoplasmic reticulum (ER), leading to ER stress, with activation of the unfolded protein response (UPR). We have previously carried out genetic studies focused on PERK (which is an eIF2α kinase that is rapidly activated in response to ER stress and leads to a repression in translation) and GADD34 (which participates in the dephosphorylation of eIF2α). We reported that mtSOD1 transgenic mice that are haploinsufficient for PERK have a significantly accelerated ALS disease, while mtSOD1 mice that are mutated for GADD34 have a remarkably ameliorated disease. Guanabenz, a centrally acting oral drug approved for the treatment of hypertension, enhances the PERK pathway by selectively inhibiting GADD34-mediated dephosphorylation of eIF2α. We have now treated G93A mtSOD1 transgenic mice with guanabenz and found a significant amelioration of disease with a delay in the onset and prolongation of the early phase of disease and survival. Guanabenz-treated G93A mice have less accumulation of mtSOD1 and an enhanced phosphorylation of eIF2α at endstage. This study further emphasizes the importance of the PERK pathway in the pathogenesis of FALS and as a therapeutic target in ALS, and identifies guanabenz as a candidate drug for the treatment of ALS patients.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-proteína gliofibrilar ácida, clon GA5, ascites fluid, clone GA5, Chemicon®