Merck
  • Development of validated determination of the eleven marker compounds in Gyejibokryeong-hwan for the quality assessment using HPLC analysis.
Development of validated determination of the eleven marker compounds in Gyejibokryeong-hwan for the quality assessment using HPLC analysis.
Archives of pharmacal research (2014-03-13)
Jung-Hoon Kim, Chang-Seob Seo, Hyeun-Kyoo Shin
RESUMEN
This study was designed for quality assessment of Gyejibokryeong-hwan (GBH) preparations using the simultaneous determination of 11 marker compounds such as gallic acid, oxypaeoniflorin, amygdalin, albiflorin, paeoniflorin, benzoic acid, coumarin, cinnamic acid, benzoylpaeoniflorin, cinnamaldehyde, and paeonol using HPLC-PDA and chemometric analysis. The correlation coefficient of linear regression was >0.9993. The LOD and LOQ values were in the ranges of 0.02-2.11 and 0.06-7.34 μg/mL, respectively. The intra-day and inter-day precision was 0.07-4.76 and 0.04-5.08 %, and the recovery range was 92.19-103.24 %, with RSD values <4.5 %. The repeatability of the 11 marker compounds was <2.5 % of the RSD values. The results of quantification indicated that there was much variation in the content of the 11 marker compounds among the GBH preparations. Amygdalin and paeoniflorin were detected in high quantities in all GBH samples, whereas some compounds such as cinnamaldehyde and paeonol were not detected in GBH samples from the Korean manufacturers. The content of marker compounds in the laboratory GBH water extract was different to the commercial preparations. Results of analyses carried out with the method indicate the need for quality control of GBH preparations.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido gálico, 97.5-102.5% (titration)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zimtaldehyd, natural, ≥95%, FG
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
trans-Cinnamaldehyde, 97%
Supelco
Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis
Supelco
Ácido gálico, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Cinnamaldehyde, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Acetonitrilo, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrilo, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Acetonitrilo, analytical standard
Sigma-Aldrich
trans-Cinnamaldehyde, FCC, FG
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrilo, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)