Merck
Aquaporin 9 expression is required for l-lactate to maintain retinal neuronal survival.
Neuroscience letters (2015-02-01)
Azusa Akashi, Akiko Miki, Akiyasu Kanamori, Makoto Nakamura
RESUMEN
Aquaporin 9 (AQP9), an aquaglyceroporin, is not only permeable to water but also to non-charged solutes, such as lactate. Lactate can be an energy source for retinal neurons. This study aimed to evaluate the effect of the downregulation of AQP9 expression on the survival rates and reactive oxygen species accumulation in RGC-5 cells cultured in a medium containing lactate. The Live/Dead assay revealed that the cell death rate of RGC-5 cells transfected with the control siRNA (siControl) was 3.65%±0.75% in the 5-mM glucose medium. The death rate was significantly increased by five-fold in the no glucose and 10-mM d-lactate media but not in the 10-mM l-lactate medium. In comparison, the death rate of cells transfected with siRNA targeting AQP9 (siAQP9) was 8.07%±1.01% in the 5-mM glucose medium, which was significantly increased by two-fold in the other medium conditions, indicating that the downregulation of AQP9 expression eliminated the prosurvival effect of l-lactate. Few RGC-5 cells transfected with siControl showed dichlorofluorescein (DCF) fluorescence when cultured in 5-mM glucose and 10-mM l-lactate media. However, approximately 70% of those showed DCF fluorescence when cultured in the no glucose and 10-mM d-lactate media. The downregulation of AQP9 significantly increased the DCF fluorescence rate to 50.44%±6.13% in the 10-mM l-lactate medium, whereas, it did not increase the rate in the other medium conditions. These results demonstrate that AQP9 expression is required for l-lactate to maintain retinal neuronal survival.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent