Merck
  • Association of Schistosoma mansoni-specific IgG and IgE antibody production and clinical schistosomiasis status in a rural area of Minas Gerais, Brazil.
Association of Schistosoma mansoni-specific IgG and IgE antibody production and clinical schistosomiasis status in a rural area of Minas Gerais, Brazil.
PloS one (2014-02-08)
Deborah Negrão-Corrêa, Juliana F Fittipaldi, José Roberto Lambertucci, Mauro Martins Teixeira, Carlos Maurício de Figueiredo Antunes, Mariângela Carneiro
RESUMEN
Studies in murine models and human populations have indicated that the collagen-rich granulomatous response against parasite eggs trapped in the liver is associated with the development of severe hepatosplenic schistosomiasis, characterized by periportal fibrosis and portal hypertension. The role of the humoral response in parasite susceptibility has been well established, but its participation in disease severity remains poorly understood. In this work, we evaluated the relationship between parasite-reactive IgE and IgG levels and schistosomiasis morbidity in infected patients with similar parasite burdens. Ninety-seven Schistosoma mansoni-infected individuals were subjected to clinical examination and abdominal ultrasound analysis. IgG reactivity and IgE concentration against Schistosoma mansoni soluble egg antigens (SEA) and adult worm antigen preparation (SWAP) were evaluated by ELISA assay. Multivariable linear regression models were used to evaluate the relationship between parasite-reactive antibodies and the co-variables investigated. The study population showed low parasite burden (median 30 eggs/g feces), constant re-infection, and signs of fibrosis was detected in more than 30% of individuals. Most infected individuals showed IgG reactivity, and the median concentrations of IgE anti-SEA and anti-SWAP antibodies were 1,870 and 1,375 ng/mL, respectively. There was no association between parasite burden and antibody response or any parameter of disease severity. However, IgG anti-SWAP level was positively associated with morbidity parameters, such as spleen size and thickness of portal vein at the entrance and secondary branch. In contrast, the data also revealed independent inverse correlations between concentration of parasite-reactive IgE and gallbladder wall thickness, a marker of fibrosis in schistosomiasis. The data indicate that IgG anti-SWAP is positively associated with severe schistosomiasis, independently of parasite burden, while high production of parasite-specific IgE is associated with mild disease in the human population. Antibody profiles are good correlates for schistosomiasis severity and could be tested as biomarkers of disease severity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%
Sigma-Aldrich
Cyanogen bromide, 99.995% trace metals basis