Merck
Increased expression of costimulatory molecules CD86 and sCTLA-4 in patients with acute lymphoblastic leukemia.
Leukemia & lymphoma (2013-11-29)
Amany Mansour, Tawfik Elkhodary, Ahmad Darwish, Mohamed Mabed
RESUMEN
We herein evaluate the role of the B7-family molecule CD86 and the Cytotoxic T-lymphocyte-associated antigen-4 (CTLA-4) as a possible immunopathogenic factors in patients with ALL. The results of 60 patients with de novo ALL were compared to 40 controls. A significant statistical difference between CD86 expression and sCTLA-4 levels in patients versus their controls has been detected. During follow up period of 28 months, patients suffered from relapse (16 patients) had significantly higher CD86 expression and sCTL-4 levels compared to those remained in complete remission (44 patients) (p = 0.005 and 0.03 respectively). Patients who died from the disease (9 patients) showed significantly higher CD 86 expression and sCTLA-4 levels than surviving patients (51 patients) (p = 0.004 and 0.01 respectively). In conclusion, the higher levels of sCTLA-4 and CD86 in B-ALL patients might be candidate parameters for poor prognosis and may serve to refine treatment stratification with intensification of therapy in those patients prone to relapse.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Mouse CTLA-4 ELISA Kit, for serum, plasma and cell culture supernatant