Merck
Nerve growth factor promotes killing of Leishmania donovani by macrophages through the induction of hydrogen peroxide.
Microbes and infection (2014-06-18)
Rieko Chiba, Yosuke Amagai, Akane Tanaka, Ken Katakura, Hiroshi Matsuda
RESUMEN
Visceral leishmaniasis is protozoonosis that occurs worldwide and still requires effective therapies with less toxicity. In this study, we examined the antileishmanial effect of nerve growth factor (NGF) using a murine infection model. NGF blocked the infection of macrophages by Leishmania donovani, which was completely cancelled by a hydrogen peroxide inhibitor. In vivo, not only did NGF show antileishmanial effects, but combination therapy of NGF and sodium stibogluconate synergistically exhibited the activity more potently than each monotherapy. These results indicate that NGF exerts antileishmanial effect by stimulating hydrogen peroxide production in macrophages and can be a novel therapy for leishmaniasis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-Glutatión reducido, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutatión reducido, ≥98.0%
Supelco
L-Glutatión reducido, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutatión reducido, BioXtra, ≥98.0%
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
L-Glutatión reducido, European Pharmacopoeia (EP) Reference Standard