Merck
Regional differences in suprathreshold intensity for bitter and umami stimuli.
Chemosensory perception (2014-12-09)
Emma L Feeney, John E Hayes
RESUMEN
The sense of taste is often referred to as a 'nutritional gatekeeper', thought to have evolved to indicate energy sources and prevent ingestion of potential toxins. Fungiform papillae are structures on the anterior tongue in which taste buds are situated. They are concentrated at the tongue's tip and they can provide a useful estimate of overall taste bud density for taste research. Some reports suggest taste perception may differ subtly across tongue regions, irrespective of FP number. Other data show an association between taste intensity perception for the bitter compound 6-n-propylthiouracil (PROP) and FP density. However, contradictions exist in the literature, with more recent, larger studies suggesting little or no association between FP number and perceived taste intensity. Much research has examined the relation between FP density and PROP perception, while other tastes have been less thoroughly studied. Here, in a cohort of mainly Caucasian individuals, aged 18-45, recruited from the campus of a large rural university, we examined regional and whole-mouth taste intensities, and FP density using an updated method of a digital still photography method first described in 2005. We found regional differences in suprathreshold intensity. Although all taste sensations were experienced all over the tongue, once again disproving the mythical tongue map, we also observed bitter and umami taste perception to be significantly greater on the posterior tongue than on the anterior tongue. In contrast, there were no regional differences observed for sweet, salty or sour tastes. The relation of FP density to whole-mouth intensity of 6-
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de potasio, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Cloruro de potasio, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Cloruro de potasio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Cloruro de potasio, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Cloruro de potasio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Cloruro de potasio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Cloruro de potasio, BioXtra, ≥99.0%
Supelco
Cloruro de potasio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cloruro de potasio, 99.999% trace metals basis
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
Cloruro de potasio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Supelco
Cloruro de potasio solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Cloruro de potasio solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Cloruro de potasio, ≥99.99% trace metals basis
Supelco
Cloruro de potasio solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Cloruro de potasio solution, 1 M KCl
Supelco
Cloruro de potasio solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Quinine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cloruro de potasio solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Cloruro de potasio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de potasio, tested according to Ph. Eur.
Supelco
L-Glutamic acid monosodium salt hydrate, analytical standard
Sigma-Aldrich
Cloruro de potasio, AnhydroBeads, −10 mesh, 99.99% trace metals basis