Merck
  • Influence of E. coli-induced prostatic inflammation on expression of androgen-responsive genes and transforming growth factor beta 1 cascade genes in rats.
Influence of E. coli-induced prostatic inflammation on expression of androgen-responsive genes and transforming growth factor beta 1 cascade genes in rats.
The Prostate (2014-12-03)
Yasuhito Funahashi, Zhou Wang, Katherine J O'Malley, Pradeep Tyagi, Donald B DeFranco, Jeffrey R Gingrich, Ryosuke Takahashi, Tsuyoshi Majima, Momokazu Gotoh, Naoki Yoshimura
RESUMEN
Prostatic inflammation is reportedly associated with the development of prostatic hyperplasia. We investigated the effects of prostatic inflammation on expression levels of androgen-responsive genes and growth factors in the rat prostate. Prostatic inflammation was induced by Escherichia coli (strain 1677) injection (0.2 ml of 1 × 10(8) CFU/ml) into the prostatic urethra of male Sprague-Dawley rats, and ventral lobes of the prostate were harvested on day 84. Rats were given 10 mg/kg celecoxib during the last month in the COX-2 inhibitor treated group. Histopathology and multiplex enzyme-linked immunosorbent assay (ELISA) for inflammation-related proteins were performed. Glandular epithelial cells and stromal regions were separately isolated using laser-capture microdissection (LCM). Real-time RT-PCR was performed to examine mRNA levels of androgen-responsive genes in the epithelium and tumor growth factor-β1 (TGF-β1) cascade genes in the stroma. Hematoxylin and eosin staining showed that mild inflammation was distributed diffusely throughout the prostate. Polymorphonuclear cells infiltrated the slightly edematous stroma, but no morphological changes were observed in the epithelium. Immunohistochemically, expression of androgen receptor and TGF-β1 in addition to IL-6 and cyclooxigenase-2 (COX-2) were enhanced in the E. coli inoculated rats. All of these factors were suppressed in the celecoxib-treated rats. Upregulation of IL-1α, IL-1β, IL-6, and RANTES in the E. coli-inoculated rats was normalized by celecoxib treatment. Significant upregulation of androgen receptor and androgen-responsive genes such as Eaf2, ELL2, FKBP5, calreticulin, and ornithine decarboxylase was observed in the LCM-dissected epithelium. Also TGF-β1 and its downstream cascade genes such as Hic-5, collagen 1, and fibronectin were upregulated significantly in the LCM-dissected stroma. The COX-2 inhibitor treatment suppressed upregulation of these genes. Prostatic inflammation changed the expression of androgen-responsive genes in the epithelium and TGF-β1 cascade genes in the stroma. Activation of TGF-β1 cascade genes in the inflamed stroma, as well as altered androgen-responsive gene expression in the epithelium, might be involved in the development of BPH. Prostate 75:381-389, 2015. © 2014 Wiley Periodicals, Inc.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Agua, suitable for HPLC
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Agua, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
Agua, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Agua, Deionized
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Supelco
Agua, suitable for ion chromatography
Sigma-Aldrich
Agua, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Etanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Agua, Molecular Biology, sterile filtered
Sigma-Aldrich
Etanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Hematoxylin
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Agua, for TOC analysis
Supelco
Etanol, standard for GC
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Agua, BioPerformance Certified
Sigma-Aldrich
Hematoxylin, certified by the BSC
Agua, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Agua, ACS reagent
Supelco
Agua, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Agua, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Etanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Etanol
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Etanol, tested according to Ph. Eur.
Sigma-Aldrich
Agua, PCR Reagent
Sigma-Aldrich
Agua, tested according to Ph. Eur.