Merck
  • A multicenter randomized trial indicates initial prednisolone treatment for childhood nephrotic syndrome for two months is not inferior to six-month treatment.
A multicenter randomized trial indicates initial prednisolone treatment for childhood nephrotic syndrome for two months is not inferior to six-month treatment.
Kidney international (2014-07-24)
Norishige Yoshikawa, Koichi Nakanishi, Mayumi Sako, Mari S Oba, Rintaro Mori, Erika Ota, Kenji Ishikura, Hiroshi Hataya, Masataka Honda, Shuichi Ito, Yuko Shima, Hiroshi Kaito, Kandai Nozu, Hidefumi Nakamura, Takashi Igarashi, Yasuo Ohashi, Kazumoto Iijima
RESUMEN
In this multicenter, open-label, randomized controlled trial, we determined whether 2-month prednisolone therapy for steroid-sensitive nephrotic syndrome was inferior or not to 6-month therapy despite significantly less steroid exposure. The primary end point was time from start of initial treatment to start of frequently relapsing nephrotic syndrome. The pre-specified non-inferiority margin was a hazard ratio of 1.3 with one-sided significance of 5%. We randomly assigned 255 children with an initial episode of steroid-sensitive nephrotic syndrome to either 2 - or 6-month treatment of which 246 were eligible for final analysis. The total prednisolone exposure counted both initial and relapse prednisolone treatment administered over 24 months. Median follow-up in months was 36.7 in the 2-month and 38.2 in the 6-month treatment group. Time to frequent relaps was similar in both groups; however, the median was reached only in the 6-month group (799 days). The hazard ratio was 0.86 (90% confidence interval, 0.64-1.16) and met the non-inferior margin. Time to first relapse was also similar in both groups: median day 242 (2-month) and 243 (6-month). Frequency and severity of adverse events were similar in both groups. Most adverse events were transient and occurred during initial or relapse therapy. Thus, 2 months of initial prednisolone therapy for steroid-sensitive nephrotic syndrome, despite less prednisolone exposure, is not inferior to 6 months of initial therapy in terms of time to onset of frequently relapsing nephrotic syndrome.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad