Merck
  • Brazilin selectively disrupts proximal IL-1 receptor signaling complex formation by targeting an IKK-upstream signaling components.
Brazilin selectively disrupts proximal IL-1 receptor signaling complex formation by targeting an IKK-upstream signaling components.
Biochemical pharmacology (2014-04-17)
Juhee Jeon, Ji Hoon Lee, Kyeong Ah Park, Hee Sun Byun, Hyunji Lee, Yoonjung Lee, Tiejun Zhang, Kidong Kang, Jeong Ho Seok, Hyun-Jung Kwon, Man-Deuk Han, Seong Wook Kang, Jang Hee Hong, Gang Min Hur
RESUMEN
The ligation of interleukin-1 receptor (IL-1R) or tumor necrosis factor receptor 1 (TNFR1) induces the recruitment of adaptor proteins and their concomitant ubiquitination to the proximal receptor signaling complex, respectively. Such are upstream signaling events of IKK that play essential roles in NF-κB activation. Thus, the discovery of a substance that would modulate the recruitment of key proximal signaling elements at the upstream level of IKK has been impending in this field of study. Here, we propose that brazilin, an active compound of Caesalpinia sappan L. (Leguminosae), is a potent NF-κB inhibitor that selectively disrupts the formation of the upstream IL-1R signaling complex. Analysis of upstream signaling events revealed that brazilin markedly abolished the IL-1β-induced polyubiquitination of IRAK1 and its interaction with IKK-γ counterpart. Notably, pretreatment of brazilin drastically interfered the recruitment of the receptor-proximal signaling components including IRAK1/4 and TRAF6 onto MyD88 in IL-1R-triggerd NF-κB activation. Interestingly, brazilin did not affect the TNF-induced RIP1 ubiquitination and the recruitment of RIP1 and TRAF2 to TNFR1, suggesting that brazilin is effective in selectively suppressing the proximal signaling complex formation of IL-1R, but not that of TNFR1. Moreover, our findings suggest that such a disruption of IL-1R-proximal complex formation by brazilin is not mediated by affecting the heterodimerization of IL-1R and IL-1RAcP. Taken together, the results suggest that the anti-IKK activity of brazilin is induced by targeting IKK upstream signaling components and subsequently disrupting proximal IL-1 receptor signaling complex formation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture