Merck
Vulnerability to sympathetic stress does not persist in takotsubo stress cardiomyopathy.
Journal of cardiac failure (2014-10-01)
Olov Collste, Mahbubul Alam, Martin Sundqvist, Jens Olson, Johan Wardell, Per Tornvall, Mats Frick
RESUMEN
Our objective was to investigate whether dobutamine stress echocardiography (DSE) could induce abnormal cardiac function in takotsubo stress cardiomyopathy (TSC) patients in a stable condition after the acute attack. This was a case-control study and a substudy of the Stockholm Myocardial Infarction With Normal Coronaries (SMINC) study. Twenty-two patients with a previous episode of TSC and 22 sex- and age-matched control subjects were recruited from the SMINC study and investigated with the use of DSE. All TSC patients had a previous normal cardiovascular magnetic resonance investigation. Tissue Doppler imaging-derived time phases of the cardiac cycle were recorded to calculate myocardial performance index (MPI) to assess ventricular function. Compared with control subjects at rest, TSC patients had a slightly but significantly higher left ventricular MPI (LV-MPI; 0.53 vs 0.59; P = .01) and as a trend higher right ventricular MPI (0.38 vs 0.47; P = .08), although during DSE these variables did not differ significantly. We found no difference in standard diastolic parameters between TSC and control subjects, but a significant higher value in LV-MPI in the TSC group at rest. However, no such difference could be demonstrated during DSE between the groups, indicating that vulnerability to sympathetic stimulation does not persist in TSC patients.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tolylene-2,4-diisocyanate, technical grade, 80%