Merck
  • Multiple reaction monitoring with multistage fragmentation (MRM3) detection enhances selectivity for LC-MS/MS analysis of plasma free metanephrines.
Multiple reaction monitoring with multistage fragmentation (MRM3) detection enhances selectivity for LC-MS/MS analysis of plasma free metanephrines.
Clinical chemistry (2015-01-22)
Michael J Wright, Rebecca L Thomas, Phoebe E Stanford, Andrea R Horvath
RESUMEN
LC-MS/MS with multiple reaction monitoring (MRM) is a powerful tool for quantifying target analytes in complex matrices. However, the technique lacks selectivity when plasma free metanephrines are measured. We propose the use of multistage fragmentation (MRM(3)) to improve the analytical selectivity of plasma free metanephrine measurement. Metanephrines were extracted from plasma with weak cation exchange solid-phase extraction before separation by hydrophilic interaction liquid chromatography. We quantified normetanephrine and metanephrine by either MRM or MRM(3) transitions m/z 166→134→79 and m/z 180→149→121, respectively. Over a 6-month period, approximately 1% (n = 21) of patient samples showed uncharacterized coeluting substances that interfered with the routine assay, resulting in an inability to report results. Quantification with MRM(3) removed these interferences and enabled measurement of the target compounds. For patient samples unaffected by interferences, Deming regression analysis demonstrated a correlation between MRM(3) and MRM methods of y = 1.00x - 0.00 nmol/L for normetanephrine and y = 0.99x + 0.03 nmol/L for metanephrine. Between the MRM(3) method and the median of all LC-MS/MS laboratories enrolled in a quality assurance program, the correlations were y = 0.97x + 0.03 nmol/L for normetanephrine and y = 1.03x - 0.04 nmol/L for metanephrine. Imprecision for the MRM(3) method was 6.2%-7.0% for normetanephrine and 6.1%-9.9% for metanephrine (n = 10). The lower limits of quantification for the MRM(3) method were 0.20 nmol/L for normetanephrine and 0.16 nmol/L for metanephrine. The use of MRM(3) technology improves the analytical selectivity of plasma free metanephrine quantification by LC-MS/MS while demonstrating sufficient analytical sensitivity and imprecision.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Cloruro de hidrógeno solution, 4.0 M in dioxane
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Supelco
Formato de amonio, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Formato de amonio, reagent grade, 97%
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ácido clorhídrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ácido clorhídrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Formato de amonio, ≥99.995% trace metals basis
Sigma-Aldrich
Cloruro de hidrógeno solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Metanol, Absolute - Acetone free
USP
Metanol, United States Pharmacopeia (USP) Reference Standard