Merck
Production of rohitukine in leaves and seeds of Dysoxylum binectariferum: an alternate renewable resource.
Pharmaceutical biology (2014-12-05)
Vidushi Mahajan, Neha Sharma, Sunil Kumar, Vikram Bhardwaj, Asif Ali, R K Khajuria, Y S Bedi, Ram A Vishwakarma, Sumit G Gandhi
RESUMEN
Rohitukine is an important precursor for the synthesis of potential anticancer drugs flavopiridol (Sanofi-Aventis) and P-276-00 (Piramal Healthcare Limited, Mumbai, India). Trunk bark of Dysoxylum binectariferum (Roxb.) Hook. f. ex Bedd. (Meliaceae) is the widely used source for isolation of rohitukine. However, removal of trunk bark threatens the survival of the tree. To investigate the amount of rohitukine accumulated in other tissues of D. binectariferum. Rohitukine standard was isolated from leaves of D. binectariferum. Its purity was ascertained using HR-MS and NMR. Crude extracts were prepared from different tissues of D. binectariferum. Rohitukine content in all the tissues was quantified by HPLC. Rohitukine accumulates in a significant amount in seeds, trunk bark, leaves, twigs, and fruits of D. binectariferum. Seeds have the highest rohitukine content (2.42%, dry weight) followed by trunk bark (1.34%, dry weight), leaves (1.064%, dry weight), twigs (0.844% dry weight), and fruits (0.4559% dry weight). Seeds and leaves of D. binectariferum could be used as alternate renewable sources for isolation of rohitukine.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O