- Distinct kinetics of viral replication, T cell infiltration, and fibrosis in three phases of myocarditis following Theiler's virus infection.
Distinct kinetics of viral replication, T cell infiltration, and fibrosis in three phases of myocarditis following Theiler's virus infection.
Cellular immunology (2014-12-03)
Fumitaka Sato, Seiichi Omura, Eiichiro Kawai, Nicholas E Martinez, Madan M Acharya, Pratap C Reddy, Ganta V Chaitanya, J Steven Alexander, Ikuo Tsunoda
PMID25460083
RESUMEN
We established a novel model of myocarditis induced with Theiler’s murine encephalomyelitis virus (TMEV), which has been used as a viral model for multiple sclerosis and seizure/epilepsy. Following TMEV infection, C3H mice developed severe myocarditis with T cell infiltration, while C57BL/6 mice had mild lesions and SJL/J mice had no inflammation in the heart. In C3H mice, myocarditis was divided into three phases: acute viral, subacute immune, and chronic fibrotic phases. Using toll-like receptor (TLR) 4-deficient C3H mice, we found that interleukin (IL)-6, IL-17, TLR4, and anti-viral immune responses were associated with myocarditis susceptibility.