Merck
Synthesis and pharmacological evaluations of 4-hydroxycoumarin derivatives as a new class of anti-Staphylococcus aureus agents.
The Journal of pharmacy and pharmacology (2015-01-06)
Zhou-peng Li, Jing Li, Di Qu, Zheng Hou, Xiao-hui Yang, Zi-dan Zhang, Yu-kun Wang, Xiao-xing Luo, Ming-kai Li
RESUMEN
Due to the increasing prevalence of drug-resistant Staphylococcus aureus infection, we develop novel 4-hydroxycoumarin derivatives as antimicrobials. The antibacterial activity of 4-hydroxycoumarin derivatives against drug-susceptive S. aureus (ATCC 29213) and methicillin-resistant S. aureus (MRSA) were evaluated using minimal inhibitory concentration (MIC) assay; the activity of favourable compound was further observed using bacterial growth curves assay and in the MRSA infection mice. Compared with dihydropyran derivatives, compound 1 as one of biscoumarins showed most potent activity with MIC values of 4-8 μg/ml and apparently inhibited the growth rate of S. aureus ATCC 29213 and USA300 strain in concentrations of both 16 and 32 mg/ml. In the mice infected with MRSA USA300, administration of 5 mg/kg compound 1 improved the animal survival rate to 66.7%, and improved the pathological change in lung tissue compared with the infection model animals. No significant cytotoxicity of compound 1 was observed on the umbilical vein endothelial cells (HUVECs) under the concentration of 800 μg/ml. Compared with the dihydropyran derivatives, biscoumarins exhibited more promising activity against both drug-sensitive and drug-resistant S. aureus, and it is efficacious in treating MRSA infections in mouse models with a favourable safety in human cells.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Dihydro-2H-pyran, 97%
Sigma-Aldrich
4-Hydroxycoumarin, 98%