Merck
Sequential dilute acid and alkali pretreatment of corn stover: sugar recovery efficiency and structural characterization.
Bioresource technology (2015-02-24)
Jae Won Lee, Ji Young Kim, Hyun Min Jang, Min Woo Lee, Jong Moon Park
RESUMEN
The objectives of this study were to explore the feasibility of applying sequential dilute acid and alkali pretreatment into the hydrolysis of corn stover and to elucidate the effects of structural changes in the biomass on its enzymatic digestibility. H2SO4 used in the first step selectively hydrolyzed 74.6-77.3% of xylan and NaOH used in the second step removed 85.9-89.4% of lignin, from the raw corn stover. Compared to single dilute acid pretreatment, the proposed combined pretreatment minimized the generation of byproducts such as acetic acid, furfural and hydroxymethylfurfural in the hydrolysates, and enhanced the enzymatic hydrolysis of the solid residue. The changes in the structural features (porosity, morphology, and crystallinity) of the solid residue were strongly correlated with the enhancement of enzymatic digestibility. The overall glucose and xylose yields finally obtained after enzymatic hydrolysis reached 89.1-97.9% and 71.0-75.9%, respectively.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido acético, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ácido acético, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ácido acético, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ácido acético solution, suitable for HPLC
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Ácido acético, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Ácido acético, United States Pharmacopeia (USP) Reference Standard
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Supelco
Ácido acético, analytical standard
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Ácido acético, ≥99.5%, FCC, FG
Sigma-Aldrich
Ácido acético, natural, ≥99.5%, FG
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Supelco
Furfural, analytical standard
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Xylose, European Pharmacopoeia (EP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Millipore
Ácido acético solution, suitable for microbiology