Merck
Clinical evaluation of the use of fiber posts and direct resin restorations for endodontically treated teeth.
The International journal of prosthodontics (2005-10-14)
Simone Grandini, Cecilia Goracci, Franklin R Tay, Romano Grandini, Marco Ferrari
RESUMEN
Restoration of root-treated teeth is routinely performed in clinical practice with a choice of therapeutic options, considering many factors to provide optimal mechanical properties, esthetics, and longevity. The aim of the present work was to present a preliminary clinical report on the use of fiber posts and direct resin composites for restoring root-treated teeth. Thirty-eight anterior and 62 posterior endodontically treated teeth were selected from 3 private prosthodontic offices. The protocol used included endodontic treatment, with translucent fiber posts (DT post) bonded to the post-space using a '1-bottle' adhesive (One-Step, Bisco) and a dual-cure resin cement (DuoLink, Bisco). Direct resin restorations were performed using a micro-hybrid resin composite (Gradia Direct, GC) and a layering technique. Both opaque dentin and enamel and translucent enamel shades were used. Patients were recalled after 6, 12, 24, and 30 months, and the restorations assessed according to predetermined clinical and radiographic criteria. These clinician-mediated evaluation methods confirmed the good clinical performance of the restorations. Restoration of endodontically treated teeth with fiber posts and direct resin composites is a treatment option, that in the short term conserves remaining tooth structure and results in good patient compliance.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Reactivos rojos para detección in situ Duolink®
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-conejo PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® Kit iniciador rojo in situ ratón o conejo
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-ratón MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Sonda para PLA® in situ Duolink® anti-conejo MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Mouse/Goat