Merck
  • Phase II Study With Epirubicin, Cisplatin, and Infusional Fluorouracil Followed by Weekly Paclitaxel With Metronomic Cyclophosphamide as a Preoperative Treatment of Triple-Negative Breast Cancer.
Phase II Study With Epirubicin, Cisplatin, and Infusional Fluorouracil Followed by Weekly Paclitaxel With Metronomic Cyclophosphamide as a Preoperative Treatment of Triple-Negative Breast Cancer.
Clinical breast cancer (2015-05-03)
Giuseppe Cancello, Vincenzo Bagnardi, Claudia Sangalli, Emilia Montagna, Silvia Dellapasqua, Andrea Sporchia, Monica Iorfida, Giuseppe Viale, Massimo Barberis, Paolo Veronesi, Alberto Luini, Mattia Intra, Aron Goldhirsch, Marco Colleoni
RESUMEN
The aggressive biological behavior and the lack of target therapy prompts the search for new therapeutic approaches for triple-negative breast cancers. We evaluated the efficacy in terms of Ki-67 variation and clinical response but also the toxicity of a neoadjuvant regimen based on metronomic principles including ECF (epidoxorubicin with cisplatin on day 1 with low-dose 5-fluorouracil in continuous infusion every 21 days for 4 courses) followed by paclitaxel (90 mg/m(2)) on day 1, 8, and 15 every 28 days for 3 courses in combination with metronomic oral cyclophosphamide 50 mg/d for 12 weeks in patients with HER2-negative breast cancer (T2-T4a-d, N0-3, M0) with estrogen receptor and progesterone receptorxa0< 10%. We enrolled 34 patients from June 2009 to May 2013. All were considered evaluable on an intention-to treat basis. The mean difference between the percentage of Ki-67 positive cells evaluated in surgical resection specimens and in pretreatment tumor core biopsy was 41% (95% confidence interval [CI], 30-51; Pxa0< .0001) for the entire population, and 22% (95% CI, 7-38; Pxa0= .0097) in patients who did not achieve pathological complete response (pCR). Responses to the treatment were obtained in 31xa0patients [91%] of the patients, and 19 patients (56%; 95% CI, 35-70) had a pCR. Stable disease was observed in 3xa0patients and none had progressive disease. Gradexa0≥ 3 hematologic adverse events included leukopenia in 9% (3 of 34), neutropenia in 38% (13 of 34), and anemia in 3% (1 of 34) of patients. Nonhematologic Gradexa0≥ 3 toxicities included only stomatitis in 1 patient. A neoadjuvant program with an ECF regimen followed by weekly paclitaxel with metronomic cyclophosphamide proved to be very effective, with high pCR rates, reduction of Ki-67, and it was associated with a low toxicity profile.