Merck
Identification and quantification of lichenysin - a possible source of food poisoning.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-09-29)
Helene Thorsen Rønning, Elisabeth Henie Madslien, Tone Normann Asp, Per Einar Granum
RESUMEN
Lichenysin produced by 53 different Bacillus licheniformis strains has been structurally examined with a qualitative liquid chromatography-tandem mass spectrometry (LC-MS/MS) method using quadrupole-time-of-flight mass spectrometry. The same lichenysin isoforms are produced from all strains, indicating that the growth conditions have a stronger influence on the lipopeptide production than the genotype. A rapid method for the quantification of lichenysin from bacterial cell cultures with LC-MS/MS after a simple methanol extraction has been refined. For the first time commercially available lichenysin has been used as calibrant, making quantification more accurate. The trueness for C15-lichenysin has been improved to 94% using matrix-matched calibration with lichenysin compared with 30% using solvent calibration with surfactin. The quantitative method was fully validated based on Commission Decision 2002/657/EC. The LOD of the method was below 1 µg g(-1) and the repeatability ranged from 10% to 16%.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)