- Identification of transforming growth factor beta family members present in bone-inductive protein purified from bovine bone.
Identification of transforming growth factor beta family members present in bone-inductive protein purified from bovine bone.
Proceedings of the National Academy of Sciences of the United States of America (1990-12-01)
A J Celeste, J A Iannazzi, R C Taylor, R M Hewick, V Rosen, E A Wang, J M Wozney
PMID2263636
RESUMEN
Characterization of the polypeptides present in bone-inductive protein extracts from bovine bone has led to the cloning of seven regulatory molecules, six of which are distantly related to transforming growth factor beta. The three human bone morphogenetic proteins (BMPs) we describe herein, BMP-5, BMP-6, and BMP-7, show extensive sequence similarity to BMP-2, a molecule that by itself is sufficient to induce de novo bone formation in vivo. The additive or synergistic contribution of these BMP-2-related molecules to the osteogenic activity associated with demineralized bone is strongly implicated by the presence of these growth factors in the most active fractions of highly purified bone extract.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bone Morphogenetic Protein 7 human, >95% (SDS-PAGE), recombinant, expressed in CHO cells, lyophilized powder, suitable for cell culture