Merck
Ki-67 labeling index affects tumor infiltration patterns of lung squamous cell carcinoma.
Molecular medicine reports (2015-10-16)
Daisuke Masuda, Ryota Masuda, Tomohiko Matsuzaki, Naoko Imamura, Naohiro Aruga, Makiko Tanaka, Sadaki Inokuchi, Hiroshi Kijima, Masayuki Iwazaki
RESUMEN
Ki-67 is a nuclear protein that is expressed during the G1, S, G2 and M phases of the mitotic cell cycle. A previous study categorized tumor infiltration patterns (INF), of which INFc indicated cancer nests exhibiting infiltrative growth and an unclear boundary between tumor tissue and surrounding healthy tissue. The present study used the Ki‑67 labeling index (Ki‑67 LI) as an indicator of cell proliferation, in order to examine the factors affecting INF in lung squamous cell carcinoma (SqCC). SqCC specimens (89) were classified into two groups: High‑grade cell proliferation (Ki‑67 LI ≥30%) and low‑grade cell proliferation (Ki‑67 LI <30%). However, a high Ki‑67 LI was significantly associated with cases that had an INFc component [INFc(+); P=0.03]. Univariate analyses indicated that INFc(+) was a predictor of venous invasion [P=0.032; odds ratio (OR), 2.615; 95% confidence interval (95% CI), 1.085‑6.305], scirrhous stromal type (P<0.001; OR, 6.462; 95% CI, 2.483‑16.817) and high Ki‑67 LI (P=0.018; OR, 12.543; 95% CI, 1.531‑102.777). Multivariate logistic analyses indicated that high Ki‑67 LI was the strongest predictor of INFc(+) (P=0.028; OR, 8.027; 95% CI, 1.248‑51.624). In conclusion, high‑grade cell proliferation activity may contribute to aggressive infiltrative growth of lung SqCC.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Peróxido de hidrógeno solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehído solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Peróxido de hidrógeno solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Sigma-Aldrich
Peróxido de hidrógeno solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Supelco
Formaldehído solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Peróxido de hidrógeno solution, 34.5-36.5%
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C