Merck
Contamination of the Hospital Environment From Potential Clostridium difficile Excretors Without Active Infection.
Infection control and hospital epidemiology (2015-04-11)
Jason S Biswas, Amita Patel, Jonathan A Otter, Esther van Kleef, Simon D Goldenberg
RESUMEN
Clostridium difficile was recovered from 33 (34%) of 98 rooms of patients who were excretors compared with 36 (49%) of 73 rooms of patients with active infection. Not all laboratory algorithms can distinguish between these 2 groups, yet both may be a significant source for ongoing transmission.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Peróxido de hidrógeno solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
Peróxido de hidrógeno solution, 34.5-36.5%
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications