Merck
Somatostatin receptor-mediated specific delivery of paclitaxel prodrugs for efficient cancer therapy.
Journal of pharmaceutical sciences (2015-03-31)
Meirong Huo, Qinnv Zhu, Qu Wu, Tingjie Yin, Lei Wang, Lifang Yin, Jianping Zhou
RESUMEN
In this study, a novel PTX prodrug, octreotide(Phe)-polyethene glycol-paclitaxel [OCT(Phe)-PEG-PTX], was successfully synthesized and used for targeted cancer therapy. A nontargeting conjugate, mPEG-PTX, was also synthesized and used as a control. Chemical structures of OCT(Phe)-PEG-PTX and mPEG-PTX were confirmed using (1) H nuclear magnetic resonance and circular dichroism. The drug contents in both the conjugates were 12.0% and 14.0%, respectively. Compared with the parent drug (PTX), OCT(Phe)-PEG-PTX, and mPEG-PTX prodrugs showed a 20,000- and 30,000-fold increase in water solubility, respectively. PTX release from mPEG-PTX and OCT(Phe)-PEG-PTX exhibited a pH-dependent profile. Moreover, compared with mPEG-PTX, OCT(Phe)-PEG-PTX exhibited significantly stronger cytotoxicity against NCI-H446 cells (SSTR overexpression) but comparable cytotoxicity against WI-38 cells (no SSTR expression). Results of confocal laser scanning microscopy revealed that the targeting prodrug labeled with fluorescence probe was selectively taken into tumor cells via SSTR-mediated endocytosis. In vivo investigation of prodrugs in nude mice bearing NCI-H446 cancer xenografts confirmed that OCT(Phe)-PEG-PTX prodrug exhibited stronger antitumor efficacy and lower systemic toxicity than mPEG-PTX and commercial Taxol. These results suggested that OCT(Phe)-PEG-PTX is a promising anticancer drug delivery system for targeted cancer therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Azul de tiazolil Bromuro de tetrazolio, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, commercial grade, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, ≥99.0% (AT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, BioXtra
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido clorhídrico solution, for amino acid analysis, ~6 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de hidrógeno solution, 3 M in cyclopentyl methyl ether (CPME)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ácido clorhídrico solution, 32 wt. % in H2O, FCC
En este momento no podemos mostrarle ni los precios ni la disponibilidad