Merck
  • Incorporating bioavailability into toxicity assessment of Cu-Ni, Cu-Cd, and Ni-Cd mixtures with the extended biotic ligand model and the WHAM-F(tox) approach.
Incorporating bioavailability into toxicity assessment of Cu-Ni, Cu-Cd, and Ni-Cd mixtures with the extended biotic ligand model and the WHAM-F(tox) approach.
Environmental science and pollution research international (2015-08-08)
Hao Qiu, Martina G Vijver, Erkai He, Yang Liu, Peng Wang, Bing Xia, Erik Smolders, Liske Versieren, Willie J G M Peijnenburg
RESUMEN
There are only a limited number of studies that have developed appropriate models which incorporate bioavailability to estimate mixture toxicity. Here, we explored the applicability of the extended biotic ligand model (BLM) and the WHAM-F(tox) approach for predicting and interpreting mixture toxicity, with the assumption that interactions between metal ions obey the BLM theory. Seedlings of lettuce Lactuca sativa were exposed to metal mixtures (Cu-Ni, Cu-Cd, and Ni-Cd) contained in hydroponic solutions for 4 days. Inhibition to root elongation was the endpoint used to quantify the toxic response. Assuming that metal ions compete with each other for binding at a single biotic ligand, the extended BLM succeeded in predicting toxicity of three mixtures to lettuce, with more than 82% of toxicity variation explained. There were no significant differences in the values of f(mix50) (i.e., the overall amounts of metal ions bound to the biotic ligand inducing 50% effect) for the three mixture combinations, showing the possibility of extrapolating these values to other binary metal combinations. The WHAM-F(tox) approach showed a similar level of precision in estimating mixture toxicity while requiring fewer parameters than the BLM-f(mix) model. External validation of the WHAM-F(tox) approach using literature data showed its applicability for other species and other mixtures. The WHAM-F(tox) model is suitable for delineating mixture effects where the extended BLM also applies. Therefore, in case of lower data availability, we recommend the lower parameterized WHAM-F(tox) as an effective approach to incorporate bioavailability in quantifying mixture toxicity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%