Merck
Toll-like receptor-4 is upregulated in plaque debris of patients with acute coronary syndrome more than Toll-like receptor-2.
Heart and vessels (2014-09-03)
Shinji Satoh, Ryoko Yada, Hiroko Inoue, Soichiro Omura, Emiko Ejima, Takahiro Mori, Katsuhiko Takenaka, Natsumi Kawamura, Kotaro Numaguchi, Etsuo Mori, Akemi Asoh, Toshihiro Nakamura, Koji Hiyamuta
RESUMEN
Atherosclerosis is a disease characterized by inflammation in the arterial wall. Atherogenesis is dependent on the innate immune response involving activation of Toll-like receptors (TLRs) and the expression of inflammatory proteins, those may lead to acute coronary syndrome (ACS). We investigated the expression level of TLR-4 in ACS, as compared with TLR-2 and patients with stable angina. Fifty-eight consecutive patients who underwent primary percutaneous coronary intervention (PCI, n = 29) because of ACS and elective PCI (n = 29) because of stable angina using a filter-device distal protection device system were prospectively analyzed. mRNA levels of TLR-2 and TLR-4 in debris containing various inflammatory tissues entrapped in the filter device were altogether analyzed using real-time PCR. There were no significant differences in age, sex distribution, between stable angina and ACS groups. TLR-4 expression levels were higher in patients with ACS than in patients with stable angina. TLR-4 might play a more important role than TLR-2 in atherogenesis, especially in ACS.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
CPNQ, ≥98% (HPLC), solid
En este momento no podemos mostrarle ni los precios ni la disponibilidad