Merck
  • 3'-Fluoro-2',3'-dideoxy-5-chlorouridine: most selective anti-HIV-1 agent among a series of new 2'- and 3'-fluorinated 2',3'-dideoxynucleoside analogues.
3'-Fluoro-2',3'-dideoxy-5-chlorouridine: most selective anti-HIV-1 agent among a series of new 2'- and 3'-fluorinated 2',3'-dideoxynucleoside analogues.
Journal of medicinal chemistry (1989-08-01)
A Van Aerschot, P Herdewijn, J Balzarini, R Pauwels, E De Clercq
RESUMEN
A series of 2'- and 3'-fluorinated 2',3'-dideoxynucleosides and 3'-azido-2',3'-dideoxynucleosides were synthesized and evaluated for their inhibitory activity against human immunodeficiency virus-1 (HIV-1) replication in MT-4 cells. Neither conversion of 3'-fluoro- or 3'-azido-2',3'-dideoxyadenosine to the corresponding inosine derivatives nor 8-bromination of 2',3'-dideoxyadenosine resulted in increased anti-HIV-1 activity. Nor did introduction of a 2'-fluorine in the erythro or threo configuration lead to improved anti-HIV-1 activity of the parent 2',3'-dideoxynucleosides. 1-(2-Fluoro-2,3-dideoxy-beta-D-threo-pentofuranosyl)cytosine and 1-(2-fluoro-2,3-dideoxy-beta-D-erythropentofuranosyl)thymine were only marginally active. However, 3'-fluoro-2',3'-dideoxyuridine (FddUrd) proved to be potent and a relatively nontoxic inhibitor of HIV-1. 5-Halogenated derivatives of FddUrd were prepared in attempts to further increase its anti-HIV potency and selectivity. Of these 5-halogenated derivatives, 3'-fluoro-2',3'-dideoxy-5-chlorouridine emerged as the most selective inhibitor of HIV-1 replication. Its selectivity index was comparable to that of azidothymidine when evaluated under the same conditions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
3′-Deoxy-3′-fluorothymidine, 97%