Merck
  • miR-181a post-transcriptionally downregulates oncogenic RalA and contributes to growth inhibition and apoptosis in chronic myelogenous leukemia (CML).
miR-181a post-transcriptionally downregulates oncogenic RalA and contributes to growth inhibition and apoptosis in chronic myelogenous leukemia (CML).
PloS one (2012-03-24)
Jia Fei, Yumin Li, Xuejiao Zhu, Xiaochuang Luo
RESUMEN
MicroRNAs (miRNAs) are a class of short RNAs that regulate gene expression through either translational repression or mRNA cleavage. miRNA-181a (miR-181a), one of the many miRNAs conserved among vertebrates, is differentially expressed in a variety of leukemia. However, its function in leukemia, particularly chronic myelogenous leukemia (CML), is poorly understood. Here we have reported the identification of miR-181a targets by combining TargetScan software prediction and expression profiling through overexpression of miR-181a mimic in leukemic K562 cells. Four overlapping genes were found to be the likely targets of miR-181a. Among the four genes, RalA is a downstream molecule of bcr-abl fusion protein in ras signaling pathway. However, its role in CML remains elusive. Luciferase reporter and Western blot assays confirmed that RalA is a direct target of miR-181a. overexpression of miR-181a effectively suppresses cell growth and induces G2-phase arrest and apoptosis partially by targeting RalA in leukemic K562 cells. Using the KEGG database combined with recent publications, downstream signaling pathway of RalA was graphed by cytoscape software. Therefore, our study is the first to report that RalA is directly regulated by miR-181a and plays an important role in CML. The approach of computational prediction combined with expression profiling might be valuable for the identification of miRNA targets in animal.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human RALA