Merck
  • α5β1 integrin induces the expression of noncartilaginous procollagen gene expression in articular chondrocytes cultured in monolayers.
α5β1 integrin induces the expression of noncartilaginous procollagen gene expression in articular chondrocytes cultured in monolayers.
Arthritis research & therapy (2013-11-30)
Nobuho Tanaka, Yasuko Ikeda, Tetsuo Yamaguchi, Hiroshi Furukawa, Hiroyuki Mitomi, Takumi Nakagawa, Shigeto Tohma, Naoshi Fukui
RESUMEN
Articular chondrocytes undergo an obvious phenotypic change when cultured in monolayers. During this change, or dedifferentiation, the expression of type I and type III procollagen is induced where normal chondrocytes express little type I and type III procollagen. In this study, we attempted to determine the mechanism(s) for the induction of such procollagen expression in dedifferentiating chondrocytes. All experiments were performed using primary-cultured human articular chondrocytes under approval of institutional review boards. Integrin(s) responsible for the induction of type I and type III procollagen expression were specified by RNAi experiments. The signal pathway(s) involved in the induction were determined by specific inhibitors and RNAi experiments. Adenovirus-mediated experiments were performed to identify a small GTPase regulating the activity of integrins in dedifferentiating chondrocytes. The effect of inhibition of integrins on dedifferentiation was investigated by experiments using echistatin, a potent disintegrin. The effect of echistatin was investigated first with monolayer-cultured chondrocytes, and then with pellet-cultured chondrocytes. In dedifferentiating chondrocytes, α5β1 integrin was found to be involved in the induction of type I and type III procollagen expression. The induction was known to be mediated by v-akt murine thymoma viral oncogene homolog (AKT) signaling. Among the three AKT isoforms, AKT1 seemed to be most involved in the signaling. Elated RAS viral (r-ras) oncogene homolog (RRAS) was considered to regulate the progression of dedifferentiation by modulating the affinity and avidity of α5β1 integrin to ligands. Echistatin inhibited dedifferentiation of monolayer-cultured chondrocytes. Furthermore, the matrix formed by pellet-cultured chondrocytes more closely resembled that of normal cartilage compared with the controls. The result of this study has shown, for the first time, that α5β1 integrin may be responsible for the induction of non-cartilaginous collagen expression in chondrocytes undergoing dedifferentiation. Again, this study has shown that the inhibition of ligand ligation to integrins may be an effective strategy to inhibit phenotypic change of cultured chondrocytes, and to improve the quality of matrix synthesized by primary cultured chondrocytes.