Merck
Development and characterization of pharmacokinetic parameters of fast-dissolving films containing levocetirizine.
Scientia pharmaceutica (2012-09-26)
Dhagla R Choudhary, Vishnu A Patel, Usmangani K Chhalotiya, Harsha V Patel, Aliasgar J Kundawala
RESUMEN
A fast-dissolving film containing levocetirizine, a non-sedative antihistamine drug, was developed using pullulan, xanthan gum, propylene glycol, and tween 80 as the base materials. The drug content of the prepared films was within an acceptable limit as prescribed by the USP. The film exhibited excellent stability for four months when stored at 40 °C and 75% humidity. In vitro dissolution studies suggested a rapid disintegration, in which most of levocetirizine (93.54 ± 3.9%) dissolved within 90 seconds after insertion into the medium. Subsequently, Sprague-Dawley rats were used to compare the pharmacokinetic properties of the film preparation administered to the oral cavity, to those with oral administration of the pure drug solution. The pharmacokinetic parameters were similar between the two groups in which AUC(0-t) (ng h/ml), AUC(0-∞) (ng h/ml) C(max) (ng/ml), T(max) (min), K(el) (h(-1)), and t(1/2) (h) of the reference were 452.033 ± 43.68, 465.78 ± 48.16, 237.16 ± 19.87, 30, 0.453 ± 0.051, and 1.536 ± 0.118, respectively, for the film formulation 447.233 ± 46.24, 458.22 ± 46.74, 233.32 ± 17.19, 30, 0.464 ± 0.060, and 1.496 ± 0.293, respectively. These results suggest that the present levocetirizine containing fast-dissolving film is likely to become one of the choices to treat different allergic conditions.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Levocetirizine dihydrochloride, ≥98% (HPLC)