Merck
Elevated NIBP/TRAPPC9 mediates tumorigenesis of cancer cells through NFκB signaling.
Oncotarget (2015-02-24)
Yonggang Zhang, Shu Liu, Hong Wang, Wensheng Yang, Fang Li, Fan Yang, Daohai Yu, Frederick V Ramsey, George P Tuszyski, Wenhui Hu
RESUMEN
Regulatory mechanisms underlying constitutive and inducible NFκB activation in cancer remain largely unknown. Here we investigated whether a novel NIK- and IKK2-binding protein (NIBP) is required for maintaining malignancy of cancer cells in an NFκB-dependent manner. Real-time polymerase chain reaction analysis of a human cancer survey tissue-scan cDNA array, immunostaining of a human frozen tumor tissue array and immunoblotting of a high-density reverse-phase cancer protein lysate array showed that NIBP is extensively expressed in most tumor tissues, particularly in breast and colon cancer. Lentivirus-mediated NIBP shRNA knockdown significantly inhibited the growth/proliferation, invasion/migration, colony formation and xenograft tumorigenesis of breast (MDA-MB-231) or colon (HCT116) cancer cells. NIBP overexpression in HCT116 cells promoted cell proliferation, migration and colony formation. Mechanistically, NIBP knockdown in cancer cells inhibited cytokine-induced activation of NFκB luciferase reporter, thus sensitizing the cells to TNFα-induced apoptosis. Endogenous NIBP bound specifically to the phosphorylated IKK2 in a TNFα-dependent manner. NIBP knockdown transiently attenuated TNFα-stimulated phosphorylation of IKK2/p65 and degradation of IκBα. In contrast, NIBP overexpression enhanced TNFα-induced NFκB activation, thus inhibiting constitutive and TNFα-induced apoptosis. Collectively, our data identified important roles of NIBP in promoting tumorigenesis via NFκΒ signaling, spotlighting NIBP as a promising target in cancer therapeutic intervention.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Roche
Pefabloc® SC PLUS, crystalline, suitable for blocking, suitable for protein purification, 0.1-1.0 mg/mL
En este momento no podemos mostrarle ni los precios ni la disponibilidad