Merck
Functionally distinct PI 3-kinase pathways regulate myelination in the peripheral nervous system.
The Journal of cell biology (2014-04-02)
Bradley A Heller, Monica Ghidinelli, Jakob Voelkl, Steven Einheber, Ryan Smith, Ethan Grund, Grant Morahan, David Chandler, Luba Kalaydjieva, Filippo Giancotti, Rosalind H King, Aniko Naray Fejes-Toth, Gerard Fejes-Toth, Maria Laura Feltri, Florian Lang, James L Salzer
RESUMEN
The PI 3-kinase (PI 3-K) signaling pathway is essential for Schwann cell myelination. Here we have characterized PI 3-K effectors activated during myelination by probing myelinating cultures and developing nerves with an antibody that recognizes phosphorylated substrates for this pathway. We identified a discrete number of phospho-proteins including the S6 ribosomal protein (S6rp), which is down-regulated at the onset of myelination, and N-myc downstream-regulated gene-1 (NDRG1), which is up-regulated strikingly with myelination. We show that type III Neuregulin1 on the axon is the primary activator of S6rp, an effector of mTORC1. In contrast, laminin-2 in the extracellular matrix (ECM), signaling through the α6β4 integrin and Sgk1 (serum and glucocorticoid-induced kinase 1), drives phosphorylation of NDRG1 in the Cajal bands of the abaxonal compartment. Unexpectedly, mice deficient in α6β4 integrin signaling or Sgk1 exhibit hypermyelination during development. These results identify functionally and spatially distinct PI 3-K pathways: an early, pro-myelinating pathway driven by axonal Neuregulin1 and a later-acting, laminin-integrin-dependent pathway that negatively regulates myelination.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-proteína básica de mielina, Chemicon®, from chicken
Sigma-Aldrich
Anti-Po Antibody, Chemicon®, from chicken