Merck
Altered Ca(2+) signaling in skeletal muscle fibers of the R6/2 mouse, a model of Huntington's disease.
The Journal of general physiology (2014-10-29)
Peter Braubach, Murat Orynbayev, Zoita Andronache, Tanja Hering, Georg Bernhard Landwehrmeyer, Katrin S Lindenberg, Werner Melzer
RESUMEN
Huntington's disease (HD) is caused by an expanded CAG trinucleotide repeat within the gene encoding the protein huntingtin. The resulting elongated glutamine (poly-Q) sequence of mutant huntingtin (mhtt) affects both central neurons and skeletal muscle. Recent reports suggest that ryanodine receptor-based Ca(2+) signaling, which is crucial for skeletal muscle excitation-contraction coupling (ECC), is changed by mhtt in HD neurons. Consequently, we searched for alterations of ECC in muscle fibers of the R6/2 mouse, a mouse model of HD. We performed fluorometric recordings of action potentials (APs) and cellular Ca(2+) transients on intact isolated toe muscle fibers (musculi interossei), and measured L-type Ca(2+) inward currents on internally dialyzed fibers under voltage-clamp conditions. Both APs and AP-triggered Ca(2+) transients showed slower kinetics in R6/2 fibers than in fibers from wild-type mice. Ca(2+) removal from the myoplasm and Ca(2+) release flux from the sarcoplasmic reticulum were characterized using a Ca(2+) binding and transport model, which indicated a significant reduction in slow Ca(2+) removal activity and Ca(2+) release flux both after APs and under voltage-clamp conditions. In addition, the voltage-clamp experiments showed a highly significant decrease in L-type Ca(2+) channel conductance. These results indicate profound changes of Ca(2+) turnover in skeletal muscle of R6/2 mice and suggest that these changes may be associated with muscle pathology in HD.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Sigma-Aldrich
Anti-Ryanodine Receptor 1 Antibody, serum, Chemicon®