Merck
  • Changes in the bound aroma profiles of 'Hayward' and 'Hort16A' kiwifruit (Actinidia spp.) during ripening and GC-olfactometry analysis.
Changes in the bound aroma profiles of 'Hayward' and 'Hort16A' kiwifruit (Actinidia spp.) during ripening and GC-olfactometry analysis.
Food chemistry (2012-12-04)
Coralia V Garcia, Ralph J Stevenson, Ross G Atkinson, Robert A Winz, Siew-Young Quek
RESUMEN
Bound volatiles are recognised as a potential source of aroma compounds in fruits. In this study, the bound volatiles of Actinidia deliciosa 'Hayward' and A. chinensis 'Hort16A' were studied at three different ripening stages. The bound volatile content tended to increase as the fruit ripened from under-ripe to ripe, and then decreased in over-ripe fruit. Glycosides of (Z)-3-hexen-1-ol and hexanol (green-note volatiles) were present in considerable amounts. β-Glucosidase activity in 'Hayward' and 'Hort16A' remained fairly constant throughout ripening. GC-olfactometry analysis of the hydrolysates of ripe 'Hayward' and 'Hort16A' revealed the presence of 2-phenylethanol, β-damascenone, vanillin and 2,5-dimethyl-4-hydroxy-3(2H)-furanone (DMHF). This is the first report of DMHF in 'Hayward' kiwifruit. For both 'Hayward' and 'Hort16A', the odour-active compounds found in the bound volatile extracts were different from those reported as contributors to the aroma of the ripe fruit, suggesting that bound volatiles are probably not significant contributors to the aroma of ripe kiwifruit.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5-Methyl-1-hexanol, 97%