Merck
RET polymorphisms might be the risk factors for thyroid cancer.
International journal of clinical and experimental pathology (2015-07-21)
Rui-Xue Huang, Fei Yang
RESUMEN
The purpose of the study is to investigate the relationship between rs1799939, rs1800858 and rs74799832 polymorphisms of RET with thyroid cancer (TC) susceptibility. Genotypes distribution of control groups were tested by Hardy-Weinberg equilibrium (HWE). Rs1799939, rs1800858 and rs74799832 polymorphisms of RET were researched in 135 patients with TC and 135 healthy people using polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP). Odds ratio (OR) with 95% confidence interval (CI) were calculated to evaluate the association between RET polymorphisms and the risk of TC by Chi-squared test. Genotypes frequencies of the control group were consistent with HWE. The frequency of genotype AA and allele A in rs1799939 were significantly higher in patients with TC than controls (OR=3.768, P=0.046; OR=1.695, P=0.035). Genotype GG and allele G of rs1800858 remarkably increased the risk of TC (OR=2.149, P=0.039; OR=1.45, P=0.039). Moreover, CC genotype and C allele in rs74799832 polymorphism was related with TC susceptibility. (OR=2.28, P=0.049; OR=1.566, P=0.049). In present result, RET rs1799939, rs1800858 and rs74799832 polymorphisms might be the risk factors for TC.