Merck
miR-93 functions as an oncomiR for the downregulation of PDCD4 in gastric carcinoma.
Scientific reports (2016-03-30)
Hongwei Liang, Feng Wang, Danping Chu, Weijie Zhang, Zhicong Liao, Zheng Fu, Xin Yan, Hao Zhu, Wen Guo, Yujing Zhang, Wenxian Guan, Xi Chen
RESUMEN
Programmed cell death 4 (PDCD4), as a tumor suppressor gene, is frequently reduced in a variety of tumors, including gastric cancer. Previous findings have indicated that PDCD4 participates in tumorigenesis through the regulation of apoptosis, but the molecular basis of this process has not been fully elucidated, and no studies have shown the upstream regulation of this gene in gastric cancer. In this study, we used bioinformatics analysis to search for miRNAs that could potentially target PDCD4 and identified miR-93 as a candidate. Moreover, we observed the inverse correlation between miR-93 and PDCD4 protein levels, but not mRNA levels, in human gastric cancer tissues. We further experimentally validated PDCD4 as the direct target of miR-93 by evaluating PDCD4 expression in gastric cancer cells after the overexpression or knockdown of miR-93. Additionally, the biological consequences of targeting PDCD4 through miR-93 were examined using cell apoptosis assays in vitro. We demonstrated that the repression of PDCD4 through miR-93 suppressed the apoptosis of gastric cancer cells. Finally, we revealed that miR-93 promoted the development of gastric tumor growth in xenograft mice by negatively regulating PDCD4. Taken together, the findings of the present study indicated the oncogenic role of miR-93 in gastric cancer tumorigenesis through targeting PDCD4, particularly in apoptosis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human PDCD4