Merck
rad-dependent response of the chk1-encoded protein kinase at the DNA damage checkpoint.
Science (New York, N.Y.) (1996-01-19)
N C Walworth, R Bernards
RESUMEN
Exposure of eukaryotic cells to agents that generate DNA damage results in transient arrest of progression through the cell cycle. In fission yeast, the DNA damage checkpoint associated with cell cycle arrest before mitosis requires the protein kinase p56chk1. DNA damage induced by ultraviolet light, gamma radiation, or a DNA-alkylating agent has now been shown to result in phosphorylation of p56chk1. This phosphorylation decreased the mobility of p56chk1 on SDS-polyacrylamide gel electrophoresis and was abolished by a mutation in the p56chk1 catalytic domain, suggesting that it might represent autophosphorylation. Phosphorylation of p56chk1 did not occur when other checkpoint genes were inactive. Thus, p56chk1 appears to function downstream of several of the known Schizosaccharomyces pombe checkpoint gene products, including that encoded by rad3+, a gene with sequence similarity to the ATM gene mutated in patients with ataxia telangiectasia. The phosphorylation of p56chk1 provides an assayable biochemical response to activation of the DNA damage checkpoint in the G2 phase of the cell cycle.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
CHK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution