Merck
Association of UHRF1 with methylated H3K9 directs the maintenance of DNA methylation.
Nature structural & molecular biology (2012-10-02)
Scott B Rothbart, Krzysztof Krajewski, Nataliya Nady, Wolfram Tempel, Sheng Xue, Aimee I Badeaux, Dalia Barsyte-Lovejoy, Jorge Y Martinez, Mark T Bedford, Stephen M Fuchs, Cheryl H Arrowsmith, Brian D Strahl
RESUMEN
A fundamental challenge in mammalian biology has been the elucidation of mechanisms linking DNA methylation and histone post-translational modifications. Human UHRF1 (ubiquitin-like PHD and RING finger domain-containing 1) has multiple domains that bind chromatin, and it is implicated genetically in the maintenance of DNA methylation. However, molecular mechanisms underlying DNA methylation regulation by UHRF1 are poorly defined. Here we show that UHRF1 association with methylated histone H3 Lys9 (H3K9) is required for DNA methylation maintenance. We further show that UHRF1 association with H3K9 methylation is insensitive to adjacent H3 S10 phosphorylation--a known mitotic 'phospho-methyl switch'. Notably, we demonstrate that UHRF1 mitotic chromatin association is necessary for DNA methylation maintenance through regulation of the stability of DNA methyltransferase-1. Collectively, our results define a previously unknown link between H3K9 methylation and the faithful epigenetic inheritance of DNA methylation, establishing a notable mitotic role for UHRF1 in this process.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
ANTI-FLAG® M2 monoclonal antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-Glutathione-S-Transferase (GST) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anticuerpo anti-etiqueta Myc, clon 9E10, clone 9E10, Upstate®, from mouse
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Anti-Histone H3 Antibody (Dimethyl K9, Phospho S10), clone 6HH3-2C5, ascites fluid, clone 6HH3-2C5, from mouse
En este momento no podemos mostrarle ni los precios ni la disponibilidad