Merck
Rnd3-induced cell rounding requires interaction with Plexin-B2.
Journal of cell science (2016-11-03)
Brad McColl, Ritu Garg, Philippe Riou, Kirsi Riento, Anne J Ridley
RESUMEN
Rnd proteins are atypical members of the Rho GTPase family that induce actin cytoskeletal reorganization and cell rounding. Rnd proteins have been reported to bind to the intracellular domain of several plexin receptors, but whether plexins contribute to the Rnd-induced rounding response is not known. Here we show that Rnd3 interacts preferentially with plexin-B2 of the three plexin-B proteins, whereas Rnd2 interacts with all three B-type plexins, and Rnd1 shows only very weak interaction with plexin-B proteins in immunoprecipitations. Plexin-B1 has been reported to act as a GAP for R-Ras and/or Rap1 proteins. We show that all three plexin-B proteins interact with R-Ras and Rap1, but Rnd proteins do not alter this interaction or R-Ras or Rap1 activity. We demonstrate that plexin-B2 promotes Rnd3-induced cell rounding and loss of stress fibres, and enhances the inhibition of HeLa cell invasion by Rnd3. We identify the amino acids in Rnd3 that are required for plexin-B2 interaction, and show that mutation of these amino acids prevents Rnd3-induced morphological changes. These results indicate that plexin-B2 is a downstream target for Rnd3, which contributes to its cellular function.
MATERIALES
Número de producto
Marca
Descripción del producto
Millipore
Gel ANTI-FLAG® M2 Affinity, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anticuerpo anti-gliceraldehído-3-fosfato deshidrogenasa, clon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-VSV-G antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human PLXNB2