Merck
  • Effect of switching from treatment with nucleos(t)ide analogs to pegylated interferon α-2a on virological and serological responses in chronic hepatitis B patients.
Effect of switching from treatment with nucleos(t)ide analogs to pegylated interferon α-2a on virological and serological responses in chronic hepatitis B patients.
World journal of gastroenterology (2016-12-29)
Li-Ting He, Xiao-Guang Ye, Xiao-Yuan Zhou
RESUMEN
To investigate the efficacy of switching to pegylated interferon-α-2a (PegIFNα-2a) treatment in nucleos(t)ide analog (NA)-treated chronic hepatitis B (CHB) responder patients. A 48-wk prospective and retrospective treatment trial of NA-treated CHB patients who had received entecavir (ETV) for at least 48 wk and had serum hepatitis B virus (HBV)-DNA < 500 IU/mL, serum hepatitis B envelope antigen (HBeAg) < 100 S/CO, serum alanine aminotransferase, and aspartate aminotransferase levels < 2 × the upper limit of normal of 40 IU/L was performed. The effects on virological and serological responses and adverse reactions to 0.5 mg daily ETV for 48 wk vs switching to PegIFNα-2a were compared. Forty-four patients were randomized to be switched from NA treatment to the PegIFNα-2a group, and 44 patients were simultaneously randomized to the ETV group. After 48 wk of therapy, the decrease in hepatitis B surface antigen (HBsAg) levels was greater in the PegIFNα-2a group than in the ETV group (3.1340 log10 IU/mL vs 3.6950 log10 IU/mL, P = 0.00). Seven patients who were anti-HBs-positive at baseline achieved HBsAg loss when switched to PegIFNα-2a (15.91% vs 0%, P = 0.018). The HBeAg serological conversion rate was higher in the PegIFNα-2a group than in the ETV group; however, the difference was not significant because of the small sample sizes (34.38% vs 21.88%, P = 0.232). In the PegIFNα-2a group, patients with HBsAg levels < 1500 IU/mL at baseline had higher HBeAg seroconversion and HBsAg loss rates at week 48 than those with HBsAg levels ≥ 1500 IU/mL (HBeAg seroconversion: 17.86% vs 62.5%, P = 0.007; HBsAg loss: 41.67% vs 6.25%, P = 0.016). Moreover, patients with HBsAg levels < 1500 IU/mL at week 24 had higher HBsAg loss rates after therapy than those with HBsAg levels ≥ 1500 IU/mL (36.84% vs 0%, P = 0.004). However, there were no statistically significant differences in HBeAg seroconversion rates (47.06% vs 25.93%, P = 0.266). NA-treated CHB patients switched to sequential PegIFNα-2a achieved highly potent treatment termination safely.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Entecavir, ≥98% (HPLC)