Merck
CoQ deficiency causes disruption of mitochondrial sulfide oxidation, a new pathomechanism associated with this syndrome.
EMBO molecular medicine (2016-11-20)
Marta Luna-Sánchez, Agustín Hidalgo-Gutiérrez, Tatjana M Hildebrandt, Julio Chaves-Serrano, Eliana Barriocanal-Casado, Ángela Santos-Fandila, Miguel Romero, Ramy Ka Sayed, Juan Duarte, Holger Prokisch, Markus Schuelke, Felix Distelmaier, Germaine Escames, Darío Acuña-Castroviejo, Luis C López
RESUMEN
Coenzyme Q (CoQ) is a key component of the mitochondrial respiratory chain, but it also has several other functions in the cellular metabolism. One of them is to function as an electron carrier in the reaction catalyzed by sulfide:quinone oxidoreductase (SQR), which catalyzes the first reaction in the hydrogen sulfide oxidation pathway. Therefore, SQR may be affected by CoQ deficiency. Using human skin fibroblasts and two mouse models with primary CoQ deficiency, we demonstrate that severe CoQ deficiency causes a reduction in SQR levels and activity, which leads to an alteration of mitochondrial sulfide metabolism. In cerebrum of Coq9
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Anti-ETHE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3