Merck
Monodisperse polystyrene foams via polymerization of foamed emulsions: structure and mechanical properties.
Physical chemistry chemical physics : PCCP (2017-02-07)
J Elsing, T Stefanov, M D Gilchrist, C Stubenrauch
RESUMEN
Foamed styrene-in-water emulsions can serve as templates for solid polystyrene foams as the pore size d
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide, 97%, powder