Merck
  • Sublytic C5b-9 Induces Glomerular Mesangial Cell Apoptosis through the Cascade Pathway of MEKK2-p38 MAPK-IRF-1-TRADD-Caspase 8 in Rat Thy-1 Nephritis.
Sublytic C5b-9 Induces Glomerular Mesangial Cell Apoptosis through the Cascade Pathway of MEKK2-p38 MAPK-IRF-1-TRADD-Caspase 8 in Rat Thy-1 Nephritis.
Journal of immunology (Baltimore, Md. : 1950) (2017-01-01)
Ganqian Zhu, Wen Qiu, Yongting Li, Chenhui Zhao, Fengxia He, Mengya Zhou, Lulu Wang, Dan Zhao, Yanlai Lu, Jing Zhang, Yu Liu, Tianyi Yu, Yingwei Wang
RESUMEN
The apoptosis of glomerular mesangial cells (GMCs) in the early phase of rat Thy-1 nephritis (Thy-1N), a model of human mesangioproliferative glomerulonephritis (MsPGN), is primarily triggered by sublytic C5b-9. However, the mechanism of GMC apoptosis induced by sublytic C5b-9 remains unclear. In this study, we demonstrate that expressions of TNFR1-associated death domain-containing protein (TRADD) and IFN regulatory factor-1 (IRF-1) were simultaneously upregulated in the renal tissue of Thy-1N rats (in vivo) and in GMCs under sublytic C5b-9 stimulation (in vitro). In vitro, TRADD was confirmed to be a downstream gene of IRF-1, because IRF-1 could bind to TRADD gene promoter to promote its transcription, leading to caspase 8 activation and GMC apoptosis. Increased phosphorylation of p38 MAPK was verified to contribute to IRF-1 and TRADD production and caspase 8 activation, as well as to GMC apoptosis induced by sublytic C5b-9. Furthermore, phosphorylation of MEK kinase 2 (MEKK2) mediated p38 MAPK activation. More importantly, three sites (Ser
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human TRADD
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP3K2