Merck
Protective role of fructokinase blockade in the pathogenesis of acute kidney injury in mice.
Nature communications (2017-02-15)
Ana Andres-Hernando, Nanxing Li, Christina Cicerchi, Shinichiro Inaba, Wei Chen, Carlos Roncal-Jimenez, Myphuong T Le, Michael F Wempe, Tamara Milagres, Takuji Ishimoto, Mehdi Fini, Takahiko Nakagawa, Richard J Johnson, Miguel A Lanaspa
RESUMEN
Acute kidney injury is associated with high mortality, especially in intensive care unit patients. The polyol pathway is a metabolic route able to convert glucose into fructose. Here we show the detrimental role of endogenous fructose production by the polyol pathway and its metabolism through fructokinase in the pathogenesis of ischaemic acute kidney injury (iAKI). Consistent with elevated urinary fructose in AKI patients, mice undergoing iAKI show significant polyol pathway activation in the kidney cortex characterized by high levels of aldose reductase, sorbitol and endogenous fructose. Wild type but not fructokinase knockout animals demonstrate severe kidney injury associated with ATP depletion, elevated uric acid, oxidative stress and inflammation. Interestingly, both the renal injury and dysfunction in wild-type mice undergoing iAKI is significantly ameliorated when exposed to luteolin, a recently discovered fructokinase inhibitor. This study demonstrates a role for fructokinase and endogenous fructose as mediators of acute renal disease.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Sigma-Aldrich
Anti-KHK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution