Merck
miR-320a functions as a suppressor for gliomas by targeting SND1 and β-catenin, and predicts the prognosis of patients.
Oncotarget (2017-02-06)
Huining Li, Lin Yu, Jing Liu, Xiuwu Bian, Cuijuan Shi, Cuiyun Sun, Xuexia Zhou, Yanjun Wen, Dan Hua, Shujun Zhao, Linlin Ren, Tongling An, Wenjun Luo, Qian Wang, Shizhu Yu
RESUMEN
miR-320a downexpression contributes to tumorigenesis in several human cancers. However, the relevance of miR-320a to prognosis, proliferation and invasion in gliomas remains unclear. In this study, we demonstrated that miR-320a expression was decreased in human glioma tissues and cell lines. Moreover, miR-320a expression was inversely correlated with glioma grades and Ki-67 index, but positively correlated with patients' survival. Contrarily, SND1 and β-catenin expressions were positively correlated with glioma grades and Ki-67 index, but inversely correlated with miR-320a expression and patients' survival. Furthermore, two subgroups with distinct prognoses in our glioma patients of different grade, IDH status, age and KPS were identified according to expression of miR-320a, SND1 or β-catenin. Cox regression showed that miR-320a and SND1 were independent predictors and β-catenin was an auxiliary predictor for patients' survival. miR-320a overexpression suppressed the G1/S phase transition, proliferation, migration and invasion of glioblastoma cells. Mechanistically, we validated SND1 and β-catenin as direct targets of miR-320a, and found that miR-320a overexpression increased SND1-inhibited tumor suppressor p21WAF1 and decreased Smad2, Smad4, MMP2, MMP7 and cyclinD1, the pivotal downstream effectors of SND1 or β-catenin. Our findings demonstrate the potential values of miR-320a, SND1 and β-catenin as prognostic biomarkers and therapeutic candidates for malignant gliomas.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human SND1