Merck
Synthetic vaccine nanoparticles target to lymph node triggering enhanced innate and adaptive antitumor immunity.
Biomaterials (2017-04-02)
Sun-Young Kim, Young-Woock Noh, Tae Heung Kang, Jung-Eun Kim, Sohyun Kim, Soong Ho Um, Doo-Byoung Oh, Yeong-Min Park, Yong Taik Lim
RESUMEN
In this study, synthetic vaccine nanoparticles (SVNPs) that efficiently targeted lymph nodes, where immune responses against foreign antigens are primed, were developed to enhance antitumor immunity. The size (20-70 nm) and surface character (amination) of poly(γ-glutamic acid)-based SVNPs were selected for effective loading and delivery (i.e., migration and retention) of model tumor antigen (OVA) and toll-like receptor 3 agonist (poly (I:C)) to immune cells in lymph nodes. Antigen-presenting cells treated with SVNP-OVA and SVNP-IC showed higher uptake of OVA and poly (I:C) and higher secretion of inflammatory cytokines (TNF-α, IL-6) and type I interferon (IFN-α, IFN-β) than those treated with OVA and poly (I:C) alone. In vivo analysis revealed higher levels of activation markers, inflammatory cytokines, and type I IFNs in the lymph nodes of mice immunized with SVNP-IC compared to those of mice in other groups. SVNP-IC-treated mice showed significantly greater in vivo natural killer cell expansion/activation (NK1.1
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cholesteryl chloroformate, 95%
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)