Merck
Potent osteogenic activity of a novel imidazobenzimidazole derivative, IBIP.
Biochemical and biophysical research communications (2017-04-19)
Su Jung Bae, Yong Ki Min, Eun Sook Hwang
RESUMEN
Bone mass is controlled by a balance between bone resorption and formation by osteoclasts and osteoblasts, respectively. An imbalance between osteoblasts and osteoclasts increases the risk of osteoporosis and fractures. Although inhibition of osteoclasts is beneficial for preventing and treating osteoporosis, enhanced bone formation through activation of osteoblast differentiation can be a more promising therapeutic approach. In this study, we attempted to isolate small molecules that promote osteoblast differentiation and found that IBIP (3-(2,3-dimethoxyphenyl)-1-[9-methyl-2-phenyl-9H-imidazo[1,2-a]benzimidazol-3-yl]-2-propen-1-one) was a potent activator of osteoblast differentiation. Upon bone morphogenetic protein-2 (BMP2) stimulation, IBIP promoted osteoblast differentiation and increased the expression of osteoblast-specific gene markers, such as osterix and alkaline phosphatase, in a dose-dependent manner. The phosphorylation of SMADs and extracellular signal-regulated kinase (ERK) increased after IBIP treatment. While enhanced SMAD phosphorylation by IBIP was abolished by a BMP inhibitor, IBIP-induced ERK phosphorylation was sustained in the presence of this inhibitor, but was decreased by an ERK kinase inhibitor. Suppression of IBIP-induced SMAD and ERK phosphorylation diminished osteoblast differentiation. Most importantly, IBIP enhanced bone formation and calcification in a BMP2-independent manner in vitro and advanced the skeletal development of zebrafish larvae in vivo. Collectively, IBIP may have beneficial effects on bone loss through potentiation of bone formation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Benzimidazole, 98%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)